The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020808.4 | NP_065859.3 | 11 - 12 | 3806 | 73 |
XM_005273211.2 | XP_005273268.1 | 13 - 14 | 4143 | 73 | |
XM_005273212.4 | XP_005273269.1 | 13 - 14 | 3950 | 73 | |
XM_005273213.4 | XP_005273270.1 | 12 - 13 | 3843 | 73 | |
XM_011544243.2 | XP_011542545.1 | 14 - 15 | 3999 | 73 | |
XM_017001896.1 | XP_016857385.1 | 13 - 14 | 3896 | 73 | |
XM_017001897.1 | XP_016857386.1 | 13 - 14 | 4143 | 73 | |
XM_017001898.1 | XP_016857387.1 | 13 - 14 | 3917 | 73 | |
GenBank mRNA | AB037810.1 | - | 11 - 12 | 3010 | 73 |
AK022852.1 | - | 3 - 4 | 406 | 73 | |
AK093191.1 | - | 5 - 6 | 942 | 73 | |
AY168879.1 | - | 11 - 12 | 3766 | 73 | |
BC013119.1 | - | 3 - 4 | 317 | 73 | |
BC082251.1 | - | 3 - 4 | 346 | 73 | |
BC150334.1 | - | 11 - 12 | 3021 | 73 |