Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIPA1L2
Entrez Gene ID
57568
Gene Name
signal induced proliferation associated 1 like 2
Gene Aliases
SPAL2
Chromosome Location
Chr.1: 232397965 - 232630491 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020808.4 NP_065859.3 7 - 8 3087 89
XM_005273211.2 XP_005273268.1 9 - 10 3424 89
XM_005273212.4 XP_005273269.1 9 - 10 3231 89
XM_005273213.4 XP_005273270.1 8 - 9 3124 89
XM_011544243.2 XP_011542545.1 10 - 11 3280 89
XM_017001896.1 XP_016857385.1 9 - 10 3177 89
XM_017001897.1 XP_016857386.1 9 - 10 3424 89
XM_017001898.1 XP_016857387.1 9 - 10 3198 89
GenBank mRNA AB037810.1 - 7 - 8 2291 89
AY168879.1 - 7 - 8 3047 89
BC150334.1 - 7 - 8 2302 89

View Full Product Details