The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138424.1 | NP_612433.1 | 9 - 10 | 1047 | 57 |
XM_005251683.4 | XP_005251740.1 | 12 - 13 | 2036 | 57 | |
XM_006716947.2 | XP_006717010.1 | 11 - 12 | 1288 | 57 | |
XM_011518190.2 | XP_011516492.1 | 12 - 13 | 2071 | 57 | |
XM_011518191.2 | XP_011516493.1 | 12 - 13 | 2070 | 57 | |
XM_011518192.2 | XP_011516494.1 | 12 - 13 | 2068 | 57 | |
XM_011518193.2 | XP_011516495.1 | 11 - 12 | 1921 | 57 | |
XM_017014240.1 | XP_016869729.1 | 6 - 7 | 727 | 57 | |
GenBank mRNA | BC010626.1 | - | 9 - 10 | 1047 | 57 |