The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138424.1 | NP_612433.1 | 13 - 14 | 1427 | 156 |
XM_005251683.4 | XP_005251740.1 | 16 - 17 | 2416 | 156 | |
XM_006716947.2 | XP_006717010.1 | 15 - 16 | 1668 | 156 | |
XM_011518190.2 | XP_011516492.1 | 16 - 17 | 2451 | 156 | |
XM_011518191.2 | XP_011516493.1 | 16 - 17 | 2450 | 156 | |
XM_017014240.1 | XP_016869729.1 | 10 - 11 | 1107 | 156 | |
GenBank mRNA | BC010626.1 | - | 13 - 14 | 1427 | 156 |