The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172223.2 | NP_001165694.1 | 2 - 3 | 498 | 69 |
NM_053005.5 | NP_443731.2 | 1 - 2 | 285 | 69 | |
GenBank mRNA | AB028173.1 | - | 1 - 2 | 234 | 69 |
AJ580639.1 | - | 1 - 2 | 182 | 69 | |
AK296658.1 | - | 2 - 3 | 465 | 69 | |
BC035317.1 | - | 1 - 2 | 66 | 69 | |
BC047291.1 | - | 1 - 2 | 222 | 69 | |
BC058934.1 | - | 1 - 2 | 231 | 69 | |
BC067785.1 | - | 1 - 2 | 246 | 69 | |
DC413532.1 | - | 2 - 3 | 437 | 69 | |
HY032250.1 | - | - | 286 | 69 |