The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005171.4 | NP_005162.1 | 2 - 3 | 398 | 69 |
XM_011538386.2 | XP_011536688.1 | 2 - 3 | 2455 | 69 | |
XM_011538387.2 | XP_011536689.1 | 2 - 3 | 128 | 69 | |
XM_017019331.1 | XP_016874820.1 | 2 - 3 | 168 | 69 | |
XM_017019332.1 | XP_016874821.1 | 2 - 3 | 262 | 69 | |
XM_017019333.1 | XP_016874822.1 | 2 - 3 | 148 | 69 | |
XM_017019334.1 | XP_016874823.1 | 2 - 3 | 174 | 69 | |
GenBank mRNA | AK313406.1 | - | 2 - 3 | 303 | 69 |
BC029619.1 | - | 2 - 3 | 112 | 69 | |
BG502726.1 | - | 2 - 3 | 131 | 69 | |
BG545632.1 | - | 2 - 3 | 326 | 69 | |
BI712653.1 | - | 2 - 3 | 287 | 69 | |
DA468394.1 | - | 2 - 3 | 398 | 69 |