Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN11
Entrez Gene ID
5010
Gene Name
claudin 11
Gene Aliases
OSP, OTM
Chromosome Location
Chr.3: 170418865 - 170434691 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185056.1 NP_001171985.1 1 - 2 95 74
GenBank mRNA AK294087.1 - 1 - 2 66 74
DA258090.1 - 1 - 2 95 74

View Full Product Details