Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAPG
Entrez Gene ID
8774
Gene Name
NSF attachment protein gamma
Gene Aliases
GAMMASNAP
Chromosome Location
Chr.18: 10525876 - 10552769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003826.2 NP_003817.1 11 - 12 1029 67
XM_011525754.2 XP_011524056.1 12 - 13 1113 67
XM_011525756.2 XP_011524058.1 9 - 10 668 67
XM_017026063.1 XP_016881552.1 7 - 8 585 67
GenBank mRNA AK126942.1 - 6 - 7 1182 67
AK294038.1 - 10 - 11 808 67
AK314495.1 - 11 - 12 868 67
BC001889.1 - 11 - 12 865 67
CR536554.1 - 11 - 12 802 67
U78107.1 - 11 - 12 875 67

View Full Product Details