The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003826.2 | NP_003817.1 | 6 - 7 | 596 | 67 |
XM_011525754.2 | XP_011524056.1 | 7 - 8 | 680 | 67 | |
XM_011525756.2 | XP_011524058.1 | 4 - 5 | 235 | 67 | |
XM_017026063.1 | XP_016881552.1 | 2 - 3 | 152 | 67 | |
GenBank mRNA | AK126942.1 | - | 1 - 2 | 749 | 67 |
AK294038.1 | - | 5 - 6 | 375 | 67 | |
AK314495.1 | - | 6 - 7 | 435 | 67 | |
BC001889.1 | - | 6 - 7 | 432 | 67 | |
CR536554.1 | - | 6 - 7 | 369 | 67 | |
U78107.1 | - | 6 - 7 | 442 | 67 |