The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015382.3 | NP_056197.3 | 11 - 12 | 2284 | 123 |
XM_005267502.2 | XP_005267559.2 | 11 - 12 | 2446 | 123 | |
XM_011536619.2 | XP_011534921.1 | 11 - 12 | 1882 | 123 | |
XM_011536621.2 | XP_011534923.1 | 11 - 12 | 1882 | 123 | |
XM_011536622.1 | XP_011534924.1 | 2 - 3 | 209 | 123 | |
XM_017021148.1 | XP_016876637.1 | 10 - 11 | 1776 | 123 | |
XM_017021149.1 | XP_016876638.1 | 10 - 11 | 1776 | 123 | |
XM_017021150.1 | XP_016876639.1 | 10 - 11 | 1776 | 123 | |
XM_017021151.1 | XP_016876640.1 | 11 - 12 | 2478 | 123 | |
XM_017021152.1 | XP_016876641.1 | 11 - 12 | 2478 | 123 | |
XM_017021153.1 | XP_016876642.1 | 11 - 12 | 1882 | 123 | |
GenBank mRNA | AB032957.2 | - | 3 - 4 | 430 | 123 |
AK095634.1 | - | 2 - 3 | 177 | 123 |