The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003028.2 | NP_003019.2 | 1 - 2 | 1289 | 107 |
GenBank mRNA | BC136581.1 | - | 1 - 2 | 1162 | 107 |
BC136582.1 | - | 1 - 2 | 1162 | 107 | |
X75342.1 | - | 1 - 2 | 1292 | 107 |