The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003028.2 | NP_003019.2 | 4 - 5 | 1797 | 105 |
GenBank mRNA | BC136581.1 | - | 4 - 5 | 1670 | 105 |
BC136582.1 | - | 4 - 5 | 1670 | 105 | |
X75342.1 | - | 4 - 5 | 1800 | 105 |