Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAZN
Entrez Gene ID
23254
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
KAZ
Chromosome Location
Chr.1: 13893387 - 15118048 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017999.2 NP_001017999.1 3 - 4 586 87
NM_001018000.3 NP_001018000.1 3 - 4 775 87
NM_001018001.2 NP_001018001.1 3 - 4 576 87
NM_015209.2 NP_056024.1 3 - 4 838 87
NM_201628.2 NP_963922.2 3 - 4 838 87
XM_005245795.4 XP_005245852.1 5 - 6 1093 87
XM_011541074.2 XP_011539376.1 5 - 6 1100 87
XM_011541075.2 XP_011539377.1 3 - 4 756 87
XM_011541076.2 XP_011539378.1 3 - 4 901 87
XM_011541077.2 XP_011539379.1 3 - 4 1880 87
XM_011541080.2 XP_011539382.1 5 - 6 1093 87
XM_017000768.1 XP_016856257.1 5 - 6 1093 87
XM_017000769.1 XP_016856258.1 5 - 6 1093 87
XM_017000770.1 XP_016856259.1 5 - 6 1093 87
XM_017000771.1 XP_016856260.1 4 - 5 761 87
XM_017000772.1 XP_016856261.1 3 - 4 440 87
GenBank mRNA AB015329.1 - 2 - 3 233 87
AY505119.1 - 4 - 5 766 87
AY505120.1 - 3 - 4 775 87
AY505121.1 - 3 - 4 576 87
AY505122.1 - 3 - 4 586 87
BC035501.2 - 3 - 4 774 87

View Full Product Details