The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017999.2 | NP_001017999.1 | 5 - 6 | 945 | 105 |
NM_001018000.3 | NP_001018000.1 | 5 - 6 | 1134 | 105 | |
NM_001018001.2 | NP_001018001.1 | 5 - 6 | 935 | 105 | |
NM_015209.2 | NP_056024.1 | 5 - 6 | 1197 | 105 | |
NM_201628.2 | NP_963922.2 | 5 - 6 | 1197 | 105 | |
XM_005245795.4 | XP_005245852.1 | 7 - 8 | 1452 | 105 | |
XM_011541074.2 | XP_011539376.1 | 7 - 8 | 1459 | 105 | |
XM_011541075.2 | XP_011539377.1 | 5 - 6 | 1115 | 105 | |
XM_011541076.2 | XP_011539378.1 | 5 - 6 | 1260 | 105 | |
XM_011541077.2 | XP_011539379.1 | 5 - 6 | 2239 | 105 | |
XM_011541080.2 | XP_011539382.1 | 7 - 8 | 1452 | 105 | |
XM_017000768.1 | XP_016856257.1 | 7 - 8 | 1452 | 105 | |
XM_017000769.1 | XP_016856258.1 | 7 - 8 | 1452 | 105 | |
XM_017000770.1 | XP_016856259.1 | 7 - 8 | 1452 | 105 | |
XM_017000771.1 | XP_016856260.1 | 6 - 7 | 1120 | 105 | |
XM_017000772.1 | XP_016856261.1 | 5 - 6 | 799 | 105 | |
GenBank mRNA | AB015329.1 | - | 4 - 5 | 592 | 105 |
AY505119.1 | - | 6 - 7 | 1125 | 105 | |
AY505120.1 | - | 5 - 6 | 1134 | 105 | |
AY505121.1 | - | 5 - 6 | 935 | 105 | |
AY505122.1 | - | 5 - 6 | 945 | 105 | |
BC035501.2 | - | 5 - 6 | 1133 | 105 |