Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAZN
Entrez Gene ID
23254
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
KAZ
Chromosome Location
Chr.1: 13893387 - 15118048 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201628.2 NP_963922.2 9 - 10 1708 62
XM_005245795.4 XP_005245852.1 11 - 12 1963 62
XM_011541074.2 XP_011539376.1 11 - 12 1970 62
XM_011541075.2 XP_011539377.1 9 - 10 1626 62
XM_011541076.2 XP_011539378.1 9 - 10 1771 62
XM_011541077.2 XP_011539379.1 9 - 10 2750 62
XM_011541080.2 XP_011539382.1 11 - 12 1963 62
XM_011541081.2 XP_011539383.1 3 - 4 450 62
GenBank mRNA AK293806.1 - 4 - 5 514 62
DR003677.1 - 4 - 5 431 62

View Full Product Details