Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 71683366 - 71769532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135820.1 NP_001129292.1 4 - 5 1572 68
NM_013390.2 NP_037522.1 4 - 5 1572 68
XM_005251869.4 XP_005251926.1 4 - 5 1205 68
GenBank mRNA AB037833.2 - 4 - 5 1165 68
AF137030.1 - 4 - 5 1180 68
AK300405.1 - 4 - 5 1201 68
BC140773.1 - 4 - 5 1074 68
BC144510.1 - 4 - 5 1074 68
BC146780.1 - 4 - 5 1165 68
BC171782.1 - 4 - 5 1074 68

View Full Product Details