Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 71683366 - 71769532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013390.2 NP_037522.1 6 - 7 1932 75
XM_005251869.4 XP_005251926.1 6 - 7 1565 75
GenBank mRNA AB037833.2 - 6 - 7 1525 75
AF137030.1 - 6 - 7 1540 75
AK300405.1 - 6 - 7 1561 75
BC144510.1 - 6 - 7 1434 75
BC146780.1 - 6 - 7 1525 75

View Full Product Details