Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 71683366 - 71769532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135820.1 NP_001129292.1 6 - 7 1913 158
NM_013390.2 NP_037522.1 7 - 8 2102 158
XM_005251869.4 XP_005251926.1 7 - 8 1735 158
GenBank mRNA AB037833.2 - 7 - 8 1695 158
AF137030.1 - 7 - 8 1710 158
AK300405.1 - 7 - 8 1731 158
BC140773.1 - 6 - 7 1415 158
BC144510.1 - 7 - 8 1604 158
BC146780.1 - 7 - 8 1695 158
BC171782.1 - 6 - 7 1416 158

View Full Product Details