The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017029911.1 | XP_016885400.1 | 1 - 2 | 113 | 63 |
GenBank mRNA | AK289677.1 | - | 10 - 11 | 1400 | 63 |
Y16355.1 | - | 10 - 11 | 1371 | 63 |