The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003611.2 | NP_003602.1 | 15 - 16 | 2017 | 87 |
XM_005274604.2 | XP_005274661.1 | 14 - 15 | 1897 | 87 | |
XM_005274606.3 | XP_005274663.1 | 16 - 17 | 2159 | 87 | |
XM_011545592.2 | XP_011543894.1 | 16 - 17 | 1988 | 87 | |
XM_011545594.2 | XP_011543896.1 | 14 - 15 | 1438 | 87 | |
XM_011545595.1 | XP_011543897.1 | 14 - 15 | 1448 | 87 | |
XM_011545597.1 | XP_011543899.1 | 16 - 17 | 2073 | 87 | |
XM_011545598.1 | XP_011543900.1 | 4 - 5 | 516 | 87 | |
XM_017029909.1 | XP_016885398.1 | 15 - 16 | 1593 | 87 | |
XM_017029910.1 | XP_016885399.1 | 16 - 17 | 1801 | 87 | |
XM_017029911.1 | XP_016885400.1 | 7 - 8 | 838 | 87 | |
GenBank mRNA | AK225847.1 | - | 14 - 15 | 1827 | 87 |
AK290354.1 | - | 15 - 16 | 2002 | 87 | |
AK297104.1 | - | 7 - 8 | 797 | 87 | |
BC096344.3 | - | 14 - 15 | 1712 | 87 | |
GU727634.1 | - | 14 - 15 | 1658 | 87 | |
Y15164.1 | - | 15 - 16 | 1969 | 87 | |
Y16355.1 | - | 15 - 16 | 2633 | 87 |