The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011516591.1 | XP_011514893.1 | 2 - 3 | 265 | 111 |
GenBank mRNA | AF412032.1 | - | 2 - 3 | 357 | 111 |
BG722364.1 | - | 2 - 3 | 357 | 111 |