The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000697.2 | NP_000688.2 | 6 - 7 | 859 | 127 |
GenBank mRNA | BC069557.1 | - | 6 - 7 | 837 | 127 |
M35418.1 | - | 6 - 7 | 847 | 127 | |
M58704.1 | - | 6 - 7 | 839 | 127 | |
M62982.1 | - | 6 - 7 | 859 | 127 |