The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293204.1 | NP_001280133.1 | 10 - 11 | 2053 | 71 |
NM_001293205.1 | NP_001280134.1 | 9 - 10 | 1855 | 71 | |
NM_005115.4 | NP_005106.2 | 11 - 12 | 2170 | 71 | |
NM_017458.3 | NP_059447.2 | 11 - 12 | 2211 | 71 | |
GenBank mRNA | AK293302.1 | - | 11 - 12 | 2065 | 71 |
AK298238.1 | - | 9 - 10 | 1829 | 71 | |
AK302052.1 | - | 11 - 12 | 1917 | 71 | |
BC015623.2 | - | 11 - 12 | 2078 | 71 | |
KJ534884.1 | - | 10 - 11 | 2018 | 71 | |
KJ534885.1 | - | 9 - 10 | 1820 | 71 | |
KJ534886.1 | - | 10 - 11 | 2018 | 71 | |
KJ535063.1 | - | 5 - 6 | 1104 | 71 | |
X79882.2 | - | 11 - 12 | 2123 | 71 |