The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293204.1 | NP_001280133.1 | 4 - 5 | 608 | 53 |
NM_001293205.1 | NP_001280134.1 | 4 - 5 | 608 | 53 | |
NM_005115.4 | NP_005106.2 | 5 - 6 | 725 | 53 | |
NM_017458.3 | NP_059447.2 | 5 - 6 | 766 | 53 | |
GenBank mRNA | AK293302.1 | - | 5 - 6 | 620 | 53 |
AK302052.1 | - | 5 - 6 | 481 | 53 | |
BC015623.2 | - | 5 - 6 | 633 | 53 | |
BQ932800.1 | - | 5 - 6 | 654 | 53 | |
KJ534884.1 | - | 4 - 5 | 573 | 53 | |
KJ534885.1 | - | 4 - 5 | 573 | 53 | |
KJ534886.1 | - | 4 - 5 | 573 | 53 | |
KJ534975.1 | - | 4 - 5 | 573 | 53 | |
KJ534976.1 | - | 4 - 5 | 573 | 53 | |
KJ534990.1 | - | 4 - 5 | 573 | 53 | |
X79882.2 | - | 5 - 6 | 678 | 53 |