Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EVPL
Entrez Gene ID
2125
Gene Name
envoplakin
Gene Aliases
EVPK
Chromosome Location
Chr.17: 76006845 - 76027452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320747.1 NP_001307676.1 16 - 17 2349 128
NM_001988.3 NP_001979.2 16 - 17 2283 128
GenBank mRNA AK290333.1 - 16 - 17 2283 128
BC126103.1 - 16 - 17 2062 128
BC126105.1 - 16 - 17 2062 128
BC143814.1 - 16 - 17 2128 128
U53786.1 - 16 - 17 2127 128

View Full Product Details