Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP6
Entrez Gene ID
9098
Gene Name
ubiquitin specific peptidase 6
Gene Aliases
HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short
Chromosome Location
Chr.17: 5116438 - 5175032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA X63547.1 - 13 - 14 2778 83

View Full Product Details