The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002253.2 | NP_002244.1 | 29 - 30 | 4148 | 103 |
GenBank mRNA | AB209901.1 | - | 29 - 30 | 4133 | 103 |
AF035121.1 | - | 29 - 30 | 4149 | 103 | |
AF063658.1 | - | 29 - 30 | 3846 | 103 | |
AK293668.1 | - | 3 - 4 | 310 | 103 | |
BC131822.1 | - | 29 - 30 | 3908 | 103 | |
L04947.1 | - | 29 - 30 | 3840 | 103 | |
X61656.1 | - | 29 - 30 | 3840 | 103 |