Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDR
Entrez Gene ID
3791
Gene Name
kinase insert domain receptor
Gene Aliases
CD309, FLK1, VEGFR, VEGFR2
Chromosome Location
Chr.4: 55078259 - 55125595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002253.2 NP_002244.1 3 - 4 659 81
GenBank mRNA AB209901.1 - 3 - 4 644 81
AF035121.1 - 3 - 4 660 81
AF063658.1 - 3 - 4 357 81
BC131822.1 - 3 - 4 419 81
EU826563.1 - 3 - 4 357 81
FJ899739.1 - 3 - 4 357 81
L04947.1 - 3 - 4 351 81
X61656.1 - 3 - 4 351 81

View Full Product Details