The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002253.2 | NP_002244.1 | 7 - 8 | 1275 | 65 |
GenBank mRNA | AB209901.1 | - | 7 - 8 | 1260 | 65 |
AF035121.1 | - | 7 - 8 | 1276 | 65 | |
AF063658.1 | - | 7 - 8 | 973 | 65 | |
BC131822.1 | - | 7 - 8 | 1035 | 65 | |
EU826563.1 | - | 7 - 8 | 973 | 65 | |
FJ899739.1 | - | 7 - 8 | 973 | 65 | |
L04947.1 | - | 7 - 8 | 967 | 65 | |
X61656.1 | - | 7 - 8 | 967 | 65 |