Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTR8
Entrez Gene ID
93973
Gene Name
ARP8 actin-related protein 8 homolog
Gene Aliases
ARP8, INO80N, hArp8
Chromosome Location
Chr.3: 53863096 - 53882202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022899.4 NP_075050.3 6 - 7 884 93
XM_005265587.4 XP_005265644.1 6 - 7 848 93
XM_011534249.2 XP_011532551.1 6 - 7 852 93
GenBank mRNA AK022996.1 - 6 - 7 834 93
AK094507.1 - 6 - 7 782 93
BC032744.1 - 2 - 3 190 93

View Full Product Details