The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022899.4 | NP_075050.3 | 8 - 9 | 1163 | 90 |
XM_005265587.4 | XP_005265644.1 | 8 - 9 | 1127 | 90 | |
XM_011534249.2 | XP_011532551.1 | 8 - 9 | 1131 | 90 | |
GenBank mRNA | AK022996.1 | - | 8 - 9 | 1113 | 90 |
AK026232.1 | - | 3 - 4 | 316 | 90 | |
AK094507.1 | - | 8 - 9 | 1061 | 90 |