The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024956.3 | NP_079232.3 | 3 - 4 | 707 | 150 |
XM_011522071.1 | XP_011520373.1 | 2 - 3 | 191 | 150 | |
XM_017022603.1 | XP_016878092.1 | 2 - 3 | 191 | 150 | |
GenBank mRNA | AK309679.1 | - | 2 - 3 | 223 | 150 |
BC009981.2 | - | 3 - 4 | 707 | 150 |