The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130918.1 | NP_001124390.1 | 4 - 5 | 510 | 75 |
XM_017024492.1 | XP_016879981.1 | 3 - 4 | 766 | 75 | |
XM_017024493.1 | XP_016879982.1 | 5 - 6 | 720 | 75 | |
XM_017024494.1 | XP_016879983.1 | 4 - 5 | 655 | 75 | |
XM_017024495.1 | XP_016879984.1 | 2 - 3 | 1176 | 75 | |
XM_017024497.1 | XP_016879986.1 | 4 - 5 | 1075 | 75 | |
GenBank mRNA | AK097392.1 | - | 4 - 5 | 422 | 75 |
AK302952.1 | - | 4 - 5 | 456 | 75 | |
AL834151.1 | - | 4 - 5 | 1076 | 75 | |
BC041368.2 | - | 3 - 4 | 1299 | 75 |