The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014371.3 | NP_055186.3 | 4 - 5 | 556 | 92 |
NR_111971.1 | 5 - 6 | 454 | 92 | ||
XM_005259854.3 | XP_005259911.2 | 5 - 6 | 361 | 92 | |
XM_011527916.2 | XP_011526218.1 | 4 - 5 | 463 | 92 | |
GenBank mRNA | AB015332.1 | - | 4 - 5 | 403 | 92 |
AB025905.1 | - | 4 - 5 | 403 | 92 | |
AB451326.1 | - | 3 - 4 | 358 | 92 | |
AB451469.1 | - | 3 - 4 | 358 | 92 | |
AF199414.1 | - | 3 - 4 | 358 | 92 | |
AJ243467.1 | - | 4 - 5 | 404 | 92 | |
AK001594.1 | - | 4 - 5 | 423 | 92 | |
AK094736.1 | - | 5 - 6 | 321 | 92 | |
AK225046.1 | - | 4 - 5 | 423 | 92 | |
AK225291.1 | - | 4 - 5 | 423 | 92 | |
BC000713.2 | - | 4 - 5 | 408 | 92 | |
CD244113.1 | - | 5 - 6 | 305 | 92 | |
DA315789.1 | - | 5 - 6 | 438 | 92 | |
DB099586.1 | - | 5 - 6 | 459 | 92 |