The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098509.1 | NP_001091979.1 | 18 - 19 | 2655 | 77 |
NM_014853.2 | NP_055668.2 | 19 - 20 | 2792 | 77 | |
XM_011524101.2 | XP_011522403.1 | 20 - 21 | 2868 | 77 | |
XM_011524102.2 | XP_011522404.1 | 20 - 21 | 2866 | 77 | |
XM_011524103.2 | XP_011522405.1 | 20 - 21 | 2867 | 77 | |
XM_011524104.2 | XP_011522406.1 | 20 - 21 | 3392 | 77 | |
XM_011524105.2 | XP_011522407.1 | 19 - 20 | 2732 | 77 | |
XM_011524106.1 | XP_011522408.1 | 19 - 20 | 2690 | 77 | |
XM_011524107.2 | XP_011522409.1 | 19 - 20 | 2686 | 77 | |
XM_011524108.1 | XP_011522410.1 | 18 - 19 | 2555 | 77 | |
XM_017025474.1 | XP_016880963.1 | 19 - 20 | 2730 | 77 | |
XM_017025475.1 | XP_016880964.1 | 19 - 20 | 2731 | 77 | |
XM_017025476.1 | XP_016880965.1 | 14 - 15 | 2027 | 77 | |
GenBank mRNA | AB007857.3 | - | 18 - 19 | 2655 | 77 |
AB275762.1 | - | 19 - 20 | 2627 | 77 | |
AB449878.1 | - | 18 - 19 | 2480 | 77 | |
AK294955.1 | - | 18 - 19 | 2547 | 77 | |
BC039204.1 | - | 19 - 20 | 2792 | 77 |