Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC5A4
Entrez Gene ID
6527
Gene Name
solute carrier family 5 member 4
Gene Aliases
DJ90G24.4, SAAT1, SGLT3
Chromosome Location
Chr.22: 32218423 - 32355057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014227.2 NP_055042.1 3 - 4 314 61
XM_006724308.3 XP_006724371.1 4 - 5 565 61
XM_011530342.2 XP_011528644.1 5 - 6 683 61
XM_011530343.2 XP_011528645.1 3 - 4 229 61
XM_011530344.2 XP_011528646.1 2 - 3 194 61
XM_017028920.1 XP_016884409.1 5 - 6 644 61
GenBank mRNA AJ133127.1 - 3 - 4 328 61

View Full Product Details