The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014227.2 | NP_055042.1 | 7 - 8 | 666 | 85 |
XM_006724308.3 | XP_006724371.1 | 8 - 9 | 917 | 85 | |
XM_011530342.2 | XP_011528644.1 | 9 - 10 | 1035 | 85 | |
XM_011530343.2 | XP_011528645.1 | 7 - 8 | 581 | 85 | |
XM_011530344.2 | XP_011528646.1 | 6 - 7 | 546 | 85 | |
XM_017028920.1 | XP_016884409.1 | 9 - 10 | 996 | 85 | |
GenBank mRNA | AJ133127.1 | - | 7 - 8 | 680 | 85 |
U41897.1 | - | 5 - 6 | 449 | 85 |