Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASC1
Entrez Gene ID
55259
Gene Name
cancer susceptibility candidate 1
Gene Aliases
LAS1, PPP1R54
Chromosome Location
Chr.12: 25107057 - 25195177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082972.2 NP_001076441.1 4 - 5 465 121
NM_001082973.2 NP_001076442.1 3 - 4 257 121
NM_001204102.2 NP_001191031.1 5 - 6 313 121
NM_001319978.1 NP_001306907.1 3 - 4 431 121
NM_018272.4 NP_060742.3 4 - 5 275 121
XM_005253406.1 XP_005253463.1 4 - 5 389 121
XM_006719101.1 XP_006719164.1 4 - 5 389 121
XM_011520723.1 XP_011519025.1 4 - 5 389 121
XM_011520726.1 XP_011519028.1 4 - 5 389 121
XM_011520727.1 XP_011519029.1 4 - 5 389 121
XM_017019560.1 XP_016875049.1 3 - 4 354 121
XM_017019561.1 XP_016875050.1 2 - 3 361 121
XM_017019562.1 XP_016875051.1 3 - 4 239 121
XM_017019563.1 XP_016875052.1 4 - 5 271 121
XM_017019564.1 XP_016875053.1 3 - 4 239 121
XM_017019565.1 XP_016875054.1 4 - 5 389 121
XM_017019566.1 XP_016875055.1 4 - 5 600 121
XM_017019568.1 XP_016875057.1 2 - 3 183 121
GenBank mRNA AK093102.1 - 4 - 5 243 121
AK301763.1 - 4 - 5 276 121
AK302235.1 - 3 - 4 221 121
AY423543.1 - 2 - 3 173 121
BC028415.1 - 4 - 5 406 121
BC031047.1 - 4 - 5 257 121
BC047415.1 - 5 - 6 311 121
BC057795.1 - 3 - 4 255 121
BC117281.1 - 1 - 2 90 121
BC117283.1 - 1 - 2 90 121
DA753279.1 - 4 - 5 303 121

View Full Product Details