The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007245.2 | NP_001007246.1 | 4 - 5 | 759 | 146 |
NM_001197079.1 | NP_001184008.1 | 3 - 4 | 649 | 146 | |
NM_001197080.1 | NP_001184009.1 | 3 - 4 | 484 | 146 | |
NM_001550.3 | NP_001541.2 | 3 - 4 | 550 | 146 | |
NR_120333.1 | 3 - 4 | 470 | 146 | ||
XM_011516142.2 | XP_011514444.1 | 3 - 4 | 545 | 146 | |
XM_011516143.2 | XP_011514445.1 | 3 - 4 | 730 | 146 | |
XM_017012151.1 | XP_016867640.1 | 3 - 4 | 797 | 146 | |
GenBank mRNA | AK225222.1 | - | 3 - 4 | 523 | 146 |
AK297509.1 | - | 3 - 4 | 295 | 146 | |
AK298894.1 | - | 3 - 4 | 290 | 146 | |
AK308066.1 | - | 4 - 5 | 734 | 146 | |
AK313211.1 | - | 4 - 5 | 759 | 146 | |
AK315967.1 | - | 3 - 4 | 482 | 146 | |
BC001272.1 | - | 3 - 4 | 503 | 146 | |
BT019889.1 | - | 3 - 4 | 289 | 146 | |
BT019890.1 | - | 3 - 4 | 289 | 146 |