The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002689.3 | NP_002680.2 | 1 - 2 | 523 | 68 |
XM_011544877.2 | XP_011543179.1 | 1 - 2 | 462 | 68 | |
XM_011544878.2 | XP_011543180.1 | 1 - 2 | 462 | 68 | |
XM_011544879.1 | XP_011543181.1 | 1 - 2 | 462 | 68 | |
XM_011544881.2 | XP_011543183.1 | 1 - 2 | 462 | 68 | |
XM_017017452.1 | XP_016872941.1 | 1 - 2 | 462 | 68 | |
GenBank mRNA | AK025315.1 | - | 1 - 2 | 386 | 68 |
AK307831.1 | - | 1 - 2 | 413 | 68 | |
BC001347.2 | - | 1 - 2 | 424 | 68 | |
BC002990.1 | - | 1 - 2 | 424 | 68 | |
BC018813.2 | - | 1 - 2 | 424 | 68 | |
BC018814.2 | - | 1 - 2 | 424 | 68 | |
CR456737.1 | - | 1 - 2 | 82 | 68 | |
DB045992.1 | - | 1 - 2 | 523 | 68 | |
L24559.1 | - | 1 - 2 | 146 | 68 |