Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN11
Entrez Gene ID
5010
Gene Name
claudin 11
Gene Aliases
OSP, OTM
Chromosome Location
Chr.3: 170418865 - 170434691 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005602.5 NP_005593.2 1 - 2 432 74
GenBank mRNA AJ245901.1 - 1 - 2 230 74
AK312923.1 - 1 - 2 429 74
BC013577.1 - 1 - 2 420 74
BT019402.1 - 1 - 2 230 74
BT019403.1 - 1 - 2 230 74
DC328248.1 - 1 - 2 432 74

View Full Product Details