Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
Chromosome Location
Chr.1: 185734526 - 186190953 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031935.2 NP_114141.2 74 - 75 11635 64
XM_011510038.2 XP_011508340.1 74 - 75 11652 64
XM_017002437.1 XP_016857926.1 63 - 64 9585 64
GenBank mRNA AF156100.1 - 74 - 75 11638 64
AJ306906.1 - 17 - 18 2520 64

View Full Product Details