Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
Chromosome Location
Chr.1: 185734526 - 186190953 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031935.2 NP_114141.2 38 - 39 6259 79
XM_011510038.2 XP_011508340.1 38 - 39 6276 79
XM_011510041.2 XP_011508343.1 38 - 39 6284 79
XM_017002437.1 XP_016857926.1 27 - 28 4209 79
GenBank mRNA AF156100.1 - 38 - 39 6259 79
AK056557.1 - 21 - 22 3358 79

View Full Product Details