Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKP1
Entrez Gene ID
5317
Gene Name
plakophilin 1
Gene Aliases
B6P
Chromosome Location
Chr.1: 201283452 - 201332993 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000299.3 NP_000290.2 4 - 5 1095 88
NM_001005337.2 NP_001005337.1 4 - 5 1095 88
GenBank mRNA BC114571.1 - 4 - 5 978 88
BC115702.1 - 4 - 5 740 88
X79293.1 - 4 - 5 1032 88
Z34974.1 - 4 - 5 1096 88

View Full Product Details