The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286445.1 | NP_001273374.1 | 20 - 21 | 3106 | 118 |
NM_014722.3 | NP_055537.2 | 21 - 22 | 3394 | 118 | |
XM_006715275.2 | XP_006715338.1 | 20 - 21 | 3323 | 118 | |
XM_006715277.1 | XP_006715340.1 | 20 - 21 | 3244 | 118 | |
XM_006715279.2 | XP_006715342.1 | 20 - 21 | 3301 | 118 | |
XM_011515007.1 | XP_011513309.1 | 21 - 22 | 3832 | 118 | |
XM_011515008.2 | XP_011513310.1 | 21 - 22 | 3163 | 118 | |
XM_011515009.1 | XP_011513311.1 | 20 - 21 | 3035 | 118 | |
XM_011515010.2 | XP_011513312.1 | 20 - 21 | 3076 | 118 | |
XM_011515012.1 | XP_011513314.1 | 21 - 22 | 3473 | 118 | |
XM_017011523.1 | XP_016867012.1 | 20 - 21 | 2998 | 118 | |
GenBank mRNA | AB002384.1 | - | 21 - 22 | 3192 | 118 |
AK299047.1 | - | 20 - 21 | 3014 | 118 |