The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242702.1 | NP_001229631.1 | 6 - 7 | 1095 | 87 |
NM_173576.2 | NP_775847.2 | 6 - 7 | 1098 | 87 | |
XM_017016105.1 | XP_016871594.1 | 6 - 7 | 1135 | 87 | |
XM_017016106.1 | XP_016871595.1 | 6 - 7 | 1326 | 87 | |
GenBank mRNA | AK125339.1 | - | 6 - 7 | 1100 | 87 |
BC036207.1 | - | 6 - 7 | 1064 | 87 |