Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD13
Entrez Gene ID
148418
Gene Name
sterile alpha motif domain containing 13
Gene Aliases
HSD-41, HSD-42
Chromosome Location
Chr.1: 84298366 - 84350798 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134663.1 NP_001128135.1 1 - 2 192 90
GenBank mRNA AK311207.1 - 1 - 2 142 90
DB459148.2 - 1 - 2 193 90

View Full Product Details