Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCD4
Entrez Gene ID
27250
Gene Name
programmed cell death 4 (neoplastic transformation inhibitor)
Gene Aliases
H731
Chromosome Location
Chr.10: 110871795 - 110900006 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145341.3 NP_663314.1 3 - 4 407 112
GenBank mRNA AK315295.1 - 3 - 4 285 112
BX647834.1 - 2 - 3 167 112

View Full Product Details