Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
Chromosome Location
Chr.1: 169511954 - 169586632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000130.4 NP_000121.2 11 - 12 1904 98
XM_017000660.1 XP_016856149.1 10 - 11 1913 98
GenBank mRNA AK226079.1 - 11 - 12 1822 98
AK291613.1 - 11 - 12 1854 98
M14335.1 - 11 - 12 1835 98
M16967.1 - 11 - 12 1849 98

View Full Product Details