The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000130.4 | NP_000121.2 | 3 - 4 | 517 | 71 |
XM_017000660.1 | XP_016856149.1 | 2 - 3 | 526 | 71 | |
GenBank mRNA | AK226079.1 | - | 3 - 4 | 435 | 71 |
AK291613.1 | - | 3 - 4 | 467 | 71 | |
AK300469.1 | - | 2 - 3 | 375 | 71 | |
AU279657.1 | - | 3 - 4 | 517 | 71 | |
M14335.1 | - | 3 - 4 | 448 | 71 | |
M16967.1 | - | 3 - 4 | 462 | 71 |